2-(7-Chloro-4-oxo-3,4-dihydroquinazolin-2(1H)-ylidene)-3-(2,4-dichlorophenyl)-3-oxopropanenitrile ≥98%

Supplier: Ambeed
Danger

Synonyms: Ciliobrevin D

A1364787-1MG A1364787-5MG
77596-928EA 45.1 USD
77596-928 77621-810
2-(7-Chloro-4-oxo-3,4-dihydroquinazolin-2(1H)-ylidene)-3-(2,4-dichlorophenyl)-3-oxopropanenitrile ≥98%
2-(7-Chloro-4-oxo-3,4-dihydroquinazolin-2(1H)-ylidene)-3-(2,4-dichlorophenyl)-3-oxopropanenitrile

Precaución: for Research Use Only

Formula: C₁₇H₈Cl₃N₃O₂
MW: 392.62 g/mol
MDL Number: MFCD32671360
CAS Number: 1370554-01-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR