DBCO-Cy3 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-[3-[1-[6-[[3-(11,12-Didehydrodibenz[b,f]azocin-5(6H)-yl)-3-oxopropyl]amino]-6-oxohexyl]-1,3-dihydro-3,3-dimethyl-5-sulpho-2H-indol-2-ylidene]-1-propen-1-yl]-3,3-dimethyl-5-sulpho-1-(3-sulphopropyl)-3H-indolium inner salt

A1364772-1MG A1364772-5MG A1364772-10MG A1364772-25MG A1364772-50MG
77621-802EA 162.94 USD
77621-802 77621-804 77867-576 77867-577 77867-578
DBCO-Cy3 ≥98%
DBCO-Cy3

Precaución: for Research Use Only

MDL Number: MFCD28505574
CAS Number: 1782950-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR