Paromomycin sulfate ≥97%

Supplier: Ambeed
Warning

A1364665-1G A1364665-5G A1364665-25G
77596-922EA 26.65 USD
77596-922 77596-924 77621-800
Paromomycin sulfate ≥97%
Paromomycin sulfate

Precaución: for Research Use Only

Formula: C₂₃H₄₅N₅O₁₄·H₂SO₄
MDL Number: MFCD00079278
CAS Number: 1263-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR