(E)-Methyl 4-aminobut-2-enoate hydrochloride ≥97%

Supplier: Ambeed
Warning

A1364513-250MG A1364513-50MG A1364513-100MG
77621-796EA 237.93 USD
77621-796 77621-798 77621-794
(E)-Methyl 4-aminobut-2-enoate hydrochloride ≥97%
(E)-Methyl 4-aminobut-2-enoate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₂
MW: 151.5914 g/mol
CAS Number: 213125-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR