(4-Amino-2-(phenylamino)thiazol-5-yl)(5-chlorothiophen-2-yl)methanone ≥98%

Supplier: Ambeed

Synonyms: JAK2-IN-6

A1364451-10MG A1364451-1MG A1364451-5MG
77596-916EA 63.55 USD
77596-916 77596-918 77596-920
(4-Amino-2-(phenylamino)thiazol-5-yl)(5-chlorothiophen-2-yl)methanone ≥98%
(4-Amino-2-(phenylamino)thiazol-5-yl)(5-chlorothiophen-2-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClN₃OS₂
MDL Number: MFCD03234340
CAS Number: 353512-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR