2-Chloro-9-phenyl-1,10-phenanthroline ≥98%

Supplier: Ambeed
Warning

A1364363-10G A1364363-1G
77596-914EA 367.98 USD
77596-914 77621-792
2-Chloro-9-phenyl-1,10-phenanthroline ≥98%
2-Chloro-9-phenyl-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₈H₁₁ClN₂
MW: 290.7463 g/mol
MDL Number: MFCD32709513
CAS Number: 1937210-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR