2-Bromo-9-phenyl-1,10-phenanthroline ≥98%

Supplier: Ambeed
Warning

A1364362-1G A1364362-250MG A1364362-5G A1364362-25G
77596-908EA 56.38 USD
77596-908 77596-910 77596-912 77621-790
2-Bromo-9-phenyl-1,10-phenanthroline ≥98%
2-Bromo-9-phenyl-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₈H₁₁BrN₂
MW: 335.1973 g/mol
MDL Number: MFCD32643418
CAS Number: 2042493-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR