Bis(4-bromophenyl)(phenyl)phosphane ≥98%

Supplier: Ambeed
Danger

A1364218-100MG A1364218-250MG A1364218-1G
77596-900EA 31.78 USD
77596-900 77596-902 77621-788
Bis(4-bromophenyl)(phenyl)phosphane ≥98%
Bis(4-bromophenyl)(phenyl)phosphane

Precaución: for Research Use Only

Formula: C₁₈H₁₃Br₂P
MW: 420.0776 g/mol
CAS Number: 84591-80-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR