3-(N-Boc-aminomethyl)-5-bromopyridine ≥98%

Supplier: Ambeed
Warning

A1364198-1G A1364198-250MG
77596-898EA 59.45 USD
77596-898 77867-567
3-(N-Boc-aminomethyl)-5-bromopyridine ≥98%
3-(N-Boc-aminomethyl)-5-bromopyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂O₂
MDL Number: MFCD10700149
CAS Number: 943722-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR