(S)-21,39-Di-tert-butyl 1-(2,5-dioxopyrrolidin-1-yl) 9,18,23-trioxo-2,5,11,14-tetraoxa-8,17,22-triazanonatriacontane-1,21,39-tricarboxylate ≥97%

Supplier: Ambeed
Warning

Synonyms: tBuO-Ste-Glu(OtBu)-AEEA-AEEA-OSU

A1363958-1G A1363958-250MG A1363958-5G
77592-576EA 205 USD
77592-576 77596-896 77621-782
(S)-21,39-Di-tert-butyl 1-(2,5-dioxopyrrolidin-1-yl) 9,18,23-trioxo-2,5,11,14-tetraoxa-8,17,22-triazanonatriacontane-1,21,39-tricarboxylate ≥97%
(S)-21,39-Di-tert-butyl 1-(2,5-dioxopyrrolidin-1-yl) 9,18,23-trioxo-2,5,11,14-tetraoxa-8,17,22-triazanonatriacontane-1,21,39-tricarboxylate

Precaución: for Research Use Only

Formula: C₄₇H₈₂N₄O₁₅
MW: 943.1718 g/mol
MDL Number: MFCD32661370
CAS Number: 1118767-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR