6-Chloro-4-methoxypyridin-2-amine ≥95%

Supplier: Ambeed
Warning

A136395-1G A136395-250MG
77611-230EA 108.15 USD
77611-230 77611-232
6-Chloro-4-methoxypyridin-2-amine ≥95%
6-Chloro-4-methoxypyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.586 g/mol
MDL Number: MFCD18415424
CAS Number: 439146-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR