(R)-5-Amino-3-(4-((5-fluoro-2-methoxybenzamido)methyl)phenyl)-1-(1,1,1-trifluoropropan-2-yl)-1H-pyrazole-4-carboxamide ≥97%

Supplier: Ambeed
Warning

Synonyms: (R)-Pirtobrutinib

A1363920-1MG A1363920-5MG
77596-894EA 33.83 USD
77596-894 77621-780
(R)-5-Amino-3-(4-((5-fluoro-2-methoxybenzamido)methyl)phenyl)-1-(1,1,1-trifluoropropan-2-yl)-1H-pyrazole-4-carboxamide ≥97%
(R)-5-Amino-3-(4-((5-fluoro-2-methoxybenzamido)methyl)phenyl)-1-(1,1,1-trifluoropropan-2-yl)-1H-pyrazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₂₁F₄N₅O₃
MW: 479.43 g/mol
CAS Number: 2101700-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR