6-(Difluoromethoxy)picolinic acid ≥95%

Supplier: Ambeed
Warning

A136367-100MG A136367-1G A136367-250MG A136367-5G
77592-574EA 20.5 USD
77592-574 77745-969 77745-970 77745-971
6-(Difluoromethoxy)picolinic acid ≥95%
6-(Difluoromethoxy)picolinic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
MDL Number: MFCD27995886
CAS Number: 1522367-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR