Bis(2-methoxyethyl) carbonate ≥97%

Supplier: Ambeed
Warning

A1363121-5G A1363121-1G
77621-776EA 456.29 USD
77621-776 77867-554
Bis(2-methoxyethyl) carbonate ≥97%
Bis(2-methoxyethyl) carbonate

Precaución: for Research Use Only

Formula: C₇H₁₄O₅
CAS Number: 626-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR