4-(Benzo[d]thiazol-2-ylthio)-3-chloroaniline 97+%

Supplier: Ambeed
Warning

Synonyms: KRAS inhibitor-9

A1362939-10MG A1362939-1MG A1362939-5MG
77596-888EA 48.18 USD
77596-888 77596-890 77596-892
4-(Benzo[d]thiazol-2-ylthio)-3-chloroaniline 97+%
4-(Benzo[d]thiazol-2-ylthio)-3-chloroaniline

Precaución: for Research Use Only

Formula: C₁₃H₉ClN₂S₂
MW: 292.8070 g/mol
MDL Number: MFCD00227023
CAS Number: 300809-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR