5-Methoxy-3-((3-((E)-2-(6-(4-methylpiperazin-1-yl)pyridin-3-yl)vinyl)-1H-indazol-6-yl)methylene)indolin-2-one dihydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: (1E)-CFI-400437 2HCl

A1362661-100MG A1362661-25MG A1362661-50MG
77621-764EA 175.1 USD
77621-764 77621-766 77621-768
5-Methoxy-3-((3-((E)-2-(6-(4-methylpiperazin-1-yl)pyridin-3-yl)vinyl)-1H-indazol-6-yl)methylene)indolin-2-one dihydrochloride ≥98%
5-Methoxy-3-((3-((E)-2-(6-(4-methylpiperazin-1-yl)pyridin-3-yl)vinyl)-1H-indazol-6-yl)methylene)indolin-2-one dihydrochloride

Precaución: for Research Use Only

Formula: C₂₉H₃₀Cl₂N₆O₂
MDL Number: MFCD30347423
CAS Number: 1247000-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR