4-((2-Acrylamidophenyl)amino)-N-(2-chloro-6-methylphenyl)-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidine-5-carboxamide ≥98%

Supplier: Ambeed
Warning

Synonyms: DGY-06-116

A1362232-100MG A1362232-25MG A1362232-50MG
77621-758EA 151.41 USD
77621-758 77621-760 77621-762
4-((2-Acrylamidophenyl)amino)-N-(2-chloro-6-methylphenyl)-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidine-5-carboxamide ≥98%
4-((2-Acrylamidophenyl)amino)-N-(2-chloro-6-methylphenyl)-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidine-5-carboxamide

Precaución: for Research Use Only

Formula: C₃₂H₃₃ClN₈O₂
MW: 597.11 g/mol
MDL Number: MFCD32899273
CAS Number: 2556836-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR