3,4,6-Trichloropyridazine ≥90%

Supplier: Ambeed
Warning

A1361492-5G A1361492-25G
77596-882EA 5.13 USD
77596-882 77621-754
3,4,6-Trichloropyridazine ≥90%
3,4,6-Trichloropyridazine

Precaución: for Research Use Only

Formula: C₄HCl₃N₂
MW: 183.4231 g/mol
MDL Number: MFCD00834957
CAS Number: 6082-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR