[(1,2,3-η)-2-Butenyl]chloro(tricyclohexylphosphine)palladium ≥97%

Supplier: Ambeed

A1360925-100MG A1360925-250MG
77592-566EA 207.05 USD
77592-566 77596-880
[(1,2,3-η)-2-Butenyl]chloro(tricyclohexylphosphine)palladium ≥97%
[(1,2,3-η)-2-Butenyl]chloro(tricyclohexylphosphine)palladium

Precaución: for Research Use Only

Formula: C₂₂H₄₀ClPPd
CAS Number: 307494-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR