2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose ≥95%

Supplier: Ambeed
Warning

Synonyms: 1,3,4,6-Tetra-O-acetyl-GlcNAc, D-Glucosamine pentaacetate, 1,3,4,6-Tetra-O-acetyl-2-(acetylamino)-2-deoxy-D-glucopyranose, (3R,4R,5S,6R)-3-Acetamido-6-(acetoxymethyl)tetrahydro-2H-pyran-2,4,5-triyl triacetate, GlcNAc tetraacetate

A136086-1G A136086-5G
77885-158EA 57 USD
77885-158 77885-159
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose ≥95%
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₁₀
MDL Number: MFCD00067367
CAS Number: 14086-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR