tert-Butyl 1-amino-3,6,9,12,15,18,21-heptaoxatetracosan-24-oate ≥98%

Supplier: Ambeed
Warning

Synonyms: Amino-PEG7-t-butyl ester

A1359832-100MG A1359832-1G
77621-720EA 76.22 USD
77621-720 77621-722
tert-Butyl 1-amino-3,6,9,12,15,18,21-heptaoxatetracosan-24-oate ≥98%
tert-Butyl 1-amino-3,6,9,12,15,18,21-heptaoxatetracosan-24-oate

Precaución: for Research Use Only

Formula: C₂₁H₄₃NO₉
MW: 453.5674 g/mol
MDL Number: MFCD32861276
CAS Number: 2428400-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR