Methyl 3-bromo-2,6-dichlorobenzoate ≥95%

Supplier: Ambeed
Warning

A135872-1G A135872-5G
77685-172EA 50 USD
77685-172 77685-173
Methyl 3-bromo-2,6-dichlorobenzoate ≥95%
Methyl 3-bromo-2,6-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrCl₂O₂
MDL Number: MFCD22572502
CAS Number: 232277-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR