2-Amino-5-fluorobenzaldehyde ≥98%

Supplier: Ambeed
Warning

A135803-1G A135803-250MG
77670-948EA 263 USD
77670-948 77670-950
2-Amino-5-fluorobenzaldehyde ≥98%
2-Amino-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD10696885
CAS Number: 146829-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR