6-Bromo-4-methyl-5-(trifluoromethyl)pyridin-2-amine ≥98%

Supplier: Ambeed
Warning

A1355880-1G A1355880-250MG A1355880-5G
77596-868EA 115.83 USD
77596-868 77621-702 77621-704
6-Bromo-4-methyl-5-(trifluoromethyl)pyridin-2-amine ≥98%
6-Bromo-4-methyl-5-(trifluoromethyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₆BrF₃N₂
MW: 255.04 g/mol
CAS Number: 2417920-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR