(4-(((2,5-Dioxopyrrolidin-1-yl)oxy)carbonyl)phenyl)boronic acid ≥98%

Supplier: Ambeed

A1355641-100MG A1355641-250MG A1355641-5G
77867-485EA 105 USD
77867-485 77867-486 77867-487
(4-(((2,5-Dioxopyrrolidin-1-yl)oxy)carbonyl)phenyl)boronic acid ≥98%
(4-(((2,5-Dioxopyrrolidin-1-yl)oxy)carbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₆
MDL Number: MFCD28955537
CAS Number: 159276-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR