Trifluoromethyl benzenesulfonate ≥98%

Supplier: Ambeed

A1355103-1G A1355103-5G
77867-478EA 38 USD
77867-478 77867-479
Trifluoromethyl benzenesulfonate ≥98%
Trifluoromethyl benzenesulfonate

Precaución: for Research Use Only

Formula: C₇H₅F₃O₃S
MDL Number: MFCD32661379
CAS Number: 1197209-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR