3,4-Dimethylpyridin-2-amine ≥97%

Supplier: Ambeed
Danger

A135446-1G A135446-100MG A135446-5G
77670-946EA 63.48 USD
77670-946 77881-071 77881-072
3,4-Dimethylpyridin-2-amine ≥97%
3,4-Dimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C7H10N2
MDL Number: MFCD02247162
CAS Number: 823-39-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR