1,1,1-Trifluoro-4-phenylbut-3-en-2-one ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A135338-100MG A135338-250MG A135338-5G
77683-196EA 88 USD
77683-196 77683-197 77683-198
1,1,1-Trifluoro-4-phenylbut-3-en-2-one ≥95%, Ambeed.Inc
1,1,1-Trifluoro-4-phenylbut-3-en-2-one

Precaución: For research use only.

Formula: C₁₀H₇F₃O
MW: 200.15 g/mol
MDL Number: MFCD00192532
CAS Number: 3108-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR