(2S,3R)-2-Amino-N-((S)-4-amino-1-oxo-1-(((3S,6S,9S,12S,15R,18S,21S)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-((R)-1-hydroxyethyl)-12-isobutyl-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl)amino)butan-2-yl)-3-hydroxybutanamide 2,2,2-trifluoroacetate ≥99%

Supplier: Ambeed
Warning

A1352723-10MG A1352723-5MG
77621-662EA 150.38 USD
77621-662 77621-664
(2S,3R)-2-Amino-N-((S)-4-amino-1-oxo-1-(((3S,6S,9S,12S,15R,18S,21S)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-((R)-1-hydroxyethyl)-12-isobutyl-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl)amino)butan-2-yl)-3-hydroxybutanamide 2,2,2-trifluoroacetate ≥99%
(2S,3R)-2-Amino-N-((S)-4-amino-1-oxo-1-(((3S,6S,9S,12S,15R,18S,21S)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-((R)-1-hydroxyethyl)-12-isobutyl-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl)amino)butan-2-yl)-3-hydroxybutanamide 2,2,

Precaución: for Research Use Only

Formula: C₄₅H₇₅F₃N₁₄O₁₃
MW: 1077.16 g/mol
CAS Number: 2220175-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR