(S)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole ≥98%

Supplier: Ambeed
Warning

A1352083-250MG A1352083-1G
77596-832EA 236.78 USD
77596-832 77621-658
(S)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole ≥98%
(S)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₃₇H₃₄NOP
CAS Number: 1362386-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR