(S)-2-(2-(Diphenylphosphanyl)phenyl)-4,5,5-triphenyl-4,5-dihydrooxazole ≥97%

Supplier: Ambeed
Warning

A1351981-250MG A1351981-1G
77596-828EA 236.78 USD
77596-828 77621-656
(S)-2-(2-(Diphenylphosphanyl)phenyl)-4,5,5-triphenyl-4,5-dihydrooxazole ≥97%
(S)-2-(2-(Diphenylphosphanyl)phenyl)-4,5,5-triphenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₃₉H₃₀NOP
MW: 559.6354 g/mol
CAS Number: 2828432-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR