2,2'-(2,2'-Dichloro-[1,1'-biphenyl]-3,3'-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) ≥98%

Supplier: Ambeed
Warning

A1351877-5G A1351877-25G
77596-822EA 102.5 USD
77596-822 77621-652
2,2'-(2,2'-Dichloro-[1,1'-biphenyl]-3,3'-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) ≥98%
2,2'-(2,2'-Dichloro-[1,1'-biphenyl]-3,3'-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

Precaución: for Research Use Only

Formula: C₂₄H₃₀B₂Cl₂O₄
MW: 475.0206 g/mol
MDL Number: MFCD32876855
CAS Number: 2248640-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR