6,6',6''-(2,4,6-Trimethylbenzene-1,3,5-triyl)tris(2-naphthoic acid) ≥97%

Supplier: Ambeed
Warning

A1351826-1G A1351826-250MG
77596-818EA 502.25 USD
77596-818 77621-650
6,6',6''-(2,4,6-Trimethylbenzene-1,3,5-triyl)tris(2-naphthoic acid) ≥97%
6,6',6''-(2,4,6-Trimethylbenzene-1,3,5-triyl)tris(2-naphthoic acid)

Precaución: for Research Use Only

Formula: C₄₂H₃₀O₆
CAS Number: 1907697-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR