Dibenzo[g,p]chrysen-2,3,6,7,10,11,14,15-octaol ≥97%

Supplier: Ambeed
Warning

A1351727-1G A1351727-250MG
77596-812EA 746.2 USD
77596-812 77596-814
Dibenzo[g,p]chrysen-2,3,6,7,10,11,14,15-octaol ≥97%
Dibenzo[g,p]chrysen-2,3,6,7,10,11,14,15-octaol

Precaución: for Research Use Only

Formula: C₂₆H₁₆O₈
CAS Number: 1246204-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR