(SP-4-1)-Nickelate(4-), [[4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]-, tetrahydrogen ≥97%

Supplier: Ambeed
Warning

A1351717-250MG A1351717-100MG
77621-640EA 177.16 USD
77621-640 77867-423
(SP-4-1)-Nickelate(4-), [[4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]-, tetrahydrogen ≥97%
(SP-4-1)-Nickelate(4-), [[4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]-, tetrahydrogen

Precaución: for Research Use Only

Formula: C₄₈H₂₈N₄NiO₈
CAS Number: 41699-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR