Hydrogen (SP-4-1)-[[4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]ferrate(4-) (4:1) ≥97%

Supplier: Ambeed
Warning

A1351716-1G A1351716-250MG A1351716-5G A1351716-100MG
77867-420EA 365 USD
77867-420 77867-421 77867-422 77867-419
Hydrogen (SP-4-1)-[[4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]ferrate(4-) (4:1) ≥97%
Hydrogen (SP-4-1)-[[4,4',4'',4'''-(21H,23H-porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]ferrate(4-) (4:1)

Precaución: for Research Use Only

Formula: C₄₈H₂₈FeN₄O₈
CAS Number: 60146-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR