Ruthenium, bis(2,2′-bipyridine-κN1,κN1′)[[2,2′-bipyridine]-5,5′-dicarboxylato(2-)-κN1,κN1′]-, dihydrochloride ≥98%

Supplier: Ambeed
Warning

A1351686-100MG A1351686-250MG A1351686-50MG A1351686-1G
77596-810EA 221.4 USD
77596-810 77621-636 77621-638 77867-418
Ruthenium, bis(2,2′-bipyridine-κN1,κN1′)[[2,2′-bipyridine]-5,5′-dicarboxylato(2-)-κN1,κN1′]-, dihydrochloride ≥98%
Ruthenium, bis(2,2′-bipyridine-κN1,κN1′)[[2,2′-bipyridine]-5,5′-dicarboxylato(2-)-κN1,κN1′]-, dihydrochloride

Precaución: for Research Use Only

Formula: C₃₂H₂₄Cl₂N₆O₄Ru
MW: 728.55 g/mol
CAS Number: 258334-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR