1,2,3,5-Tetrachloro-4-fluorobenzene 98+%

Supplier: Ambeed
Danger

A1350170-1G A1350170-250MG A1350170-5G
77867-414EA 32 USD
77867-414 77867-415 77867-416
1,2,3,5-Tetrachloro-4-fluorobenzene 98+%
1,2,3,5-Tetrachloro-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₆HCl₄F
MDL Number: MFCD22053143
CAS Number: 319-95-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR