(+/-)-exo-6-Hydroxytropinone ≥95%

Supplier: Ambeed
Warning

Synonyms: exo-6-Hydroxytropinone, (1R,5R,6S)-rel-6-Hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-one, (+/-)-exo-6-Hydroxy-3-tropanone, (+/-)-exo-6-Hydroxy-8-methyl-8-azabicyclo[3.2.1]octan-3-one

A134871-25G A134871-1G A134871-5G
77592-548EA 511.48 USD
77592-548 77624-128 77624-130
(+/-)-exo-6-Hydroxytropinone ≥95%
(+/-)-exo-6-Hydroxytropinone

Precaución: for Research Use Only

Formula: C₈H₁₃NO₂
MW: 155.20 g/mol
MDL Number: MFCD00135648
CAS Number: 5932-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR