4-(5-(Methylsulfonyl)-1,3,4-oxadiazol-2-yl)aniline ≥97%

Supplier: Ambeed
Warning

A134683-100MG A134683-250MG
77747-962EA 323 USD
77747-962 77747-963
4-(5-(Methylsulfonyl)-1,3,4-oxadiazol-2-yl)aniline ≥97%
4-(5-(Methylsulfonyl)-1,3,4-oxadiazol-2-yl)aniline

Precaución: for Research Use Only

Formula: C₉H₉N₃O₃S
MW: 239.25 g/mol
MDL Number: MFCD31692614
CAS Number: 1858267-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR