tert-Butyl (5-aminopyridin-2-yl)(methyl)carbamate ≥97%

Supplier: Ambeed
Warning

A134670-1G A134670-250MG
77883-085EA 55 USD
77883-085 77883-086
tert-Butyl (5-aminopyridin-2-yl)(methyl)carbamate ≥97%
tert-Butyl (5-aminopyridin-2-yl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃O₂
MDL Number: MFCD11215255
CAS Number: 1039055-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR