Methyl 4-(piperidin-4-ylmethyl)benzoate hydrochloride ≥95%

Supplier: Ambeed
Warning

A134322-100MG A134322-250MG
77745-956EA 50 USD
77745-956 77745-957
Methyl 4-(piperidin-4-ylmethyl)benzoate hydrochloride ≥95%
Methyl 4-(piperidin-4-ylmethyl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₀ClNO₂
MW: 269.77 g/mol
MDL Number: MFCD02179177
CAS Number: 333986-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR