Benzyl (3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate ≥95%

Supplier: Ambeed

A134239-5G A134239-1G A134239-250MG
77889-914EA 34 USD
77889-914 77889-912 77889-913
Benzyl (3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate ≥95%
Benzyl (3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₀H₂₃BFNO₄
MW: 371.21 g/mol
MDL Number: MFCD11054035
CAS Number: 1218791-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR