2,2-Bis((methoxycarbonyl)amino)acetic acid ≥97%

Supplier: Ambeed
Warning

A134160-100MG A134160-1G A134160-250MG A134160-5G
77889-907EA 20 USD
77889-907 77889-908 77889-909 77889-910
2,2-Bis((methoxycarbonyl)amino)acetic acid ≥97%
2,2-Bis((methoxycarbonyl)amino)acetic acid

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O₆
MDL Number: MFCD10699144
CAS Number: 110599-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR