3-Amino-N-(2-bromo-4,6-difluorophenyl)-6,7-dihydro-5H-cyclopenta[b]thieno[3,2-e]pyridine-2-carboxamide ≥98%

Supplier: Ambeed
Warning

A1340310-1MG A1340310-10MG A1340310-5MG A1340310-25MG A1340310-50MG
77596-806EA 53.3 USD
77596-806 77621-618 77621-620 77867-393 77867-394
3-Amino-N-(2-bromo-4,6-difluorophenyl)-6,7-dihydro-5H-cyclopenta[b]thieno[3,2-e]pyridine-2-carboxamide ≥98%
3-Amino-N-(2-bromo-4,6-difluorophenyl)-6,7-dihydro-5H-cyclopenta[b]thieno[3,2-e]pyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₁₂BrF₂N₃OS
MDL Number: MFCD03081557
CAS Number: 497061-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR