576/589 NHS Ester ≥97%

Supplier: Ambeed
Warning

A1339770-25MG A1339770-50MG A1339770-100MG A1339770-250MG
77596-796EA 402.83 USD
77596-796 77596-798 77621-608 77621-610
576/589 NHS Ester ≥97%
576/589 NHS Ester

Precaución: for Research Use Only

MDL Number: MFCD32858277
CAS Number: 201998-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR