4-Isothiocyanatophenyl α-D-mannopyranoside ≥97%
Supplier: Ambeed
|
Danger
|
Synonyms:
(2R,3S,4S,5S,6R)-2-(Hydroxymethyl)-6-(4-isothiocyanatophenoxy)tetrahydro-2H-pyran-3,4,5-triol, α-D-Mannopyranosyl 4-phenylisothiocyanate
A1339416-100MG
A1339416-1G
A1339416-250MG
A1339416-25MG
A1339416-50MG
77621-588EA
283.25
USD
77621-588
77621-590
77621-592
77867-370
77867-371
4-Isothiocyanatophenyl α-D-mannopyranoside ≥97%
4-Isothiocyanatophenyl α-D-mannopyranoside
Precaución: for Research Use Only
Formula:
C₁₃H₁₅NO₆S MW: 313.326 g/mol |
MDL Number:
MFCD00070374 CAS Number: 96345-79-8 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...