tert-Butyl 5-oxo-2-azaspiro[3.4]octane-2-carboxylate ≥97%

Supplier: Ambeed
Warning

A133710-1G A133710-250MG
77685-154EA 580 USD
77685-154 77685-155
tert-Butyl 5-oxo-2-azaspiro[3.4]octane-2-carboxylate ≥97%
tert-Butyl 5-oxo-2-azaspiro[3.4]octane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD28155163
CAS Number: 1581683-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR