Fmoc-D-cyclopropylalanine ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-β-Cyclopropyl-Ala-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopropylpropanoic acid, Fmoc-β-cyclopropyl-D-Ala-OH

A133395-5G A133395-1G A133395-250MG
77592-536EA 137.35 USD
77592-536 77670-898 77881-060
Fmoc-D-cyclopropylalanine ≥95%
Fmoc-D-cyclopropylalanine

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD02259484
CAS Number: 170642-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR