Mupirocin ≥98%

Supplier: Ambeed
Warning

Synonyms: 9-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoic acid

A133111-1MG A133111-50MG
77592-530EA 5.13 USD
77592-530 77592-532
Mupirocin ≥98%
Mupirocin

Precaución: for Research Use Only

Formula: C₂₆H₄₄O₉
MW: 500.629 g/mol
MDL Number: MFCD01711620
CAS Number: 12650-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR